4-Phenoxybenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS22116G100
AS22116G25
ACTIAS22116G100EA
0
CHF
ACTIAS22116G100
ACTIAS22116G25
4-Phenoxybenzoesäure
4-Phenoxybenzoesäure
Formel:
C₁₃H₁₀O₃ MW: 214,22 g/mol |
MDL Number:
MFCD00002539 CAS-Nummer: 2215-77-2 EINECS: 218-682-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...