3-Hydroxy-p-anissäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS20109G100 AS20109G25
ACTIAS20109G100EA 0 CHF
ACTIAS20109G100 ACTIAS20109G25
3-Hydroxy-p-anissäure
3-Hydroxy-p-anissäure
Formel: C₈H₈O₄
Siedepunkt: 327 °C (1013 hPa)
Melting Pt: 258…259 °C
MDL Number: MFCD00002507
CAS-Nummer: 645-08-9
EINECS: 211-430-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR