6-Nitro-m-anissäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Methoxy-2-nitrobenzoic acid

AS20259G100 AS20259G25
ACTIAS20259G100EA 0 CHF
ACTIAS20259G100 ACTIAS20259G25
6-Nitro-m-anissäure
6-Nitro-m-anissäure
Formel: C₈H₇NO₅
MW: 197,15 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00151836
CAS-Nummer: 1882-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR