2-Nitro-m-anissäure
Lieferant: ACTIVATE SCIENTIFIC
AS34998G1
AS34998G5
ACTIAS34998G1EA
0
CHF
ACTIAS34998G1
ACTIAS34998G5
2-Nitro-m-anissäure
2-Nitro-m-anissäure
Formel:
C₈H₇NO₅ Siedepunkt: 388 °C (1013 hPa) Melting Pt: 174 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00007143 CAS-Nummer: 4920-80-3 EINECS: 225-549-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...