2-Methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS21933G100 AS21933G25
ACTIAS21933G100EA 0 CHF
ACTIAS21933G100 ACTIAS21933G25
2-Methoxybenzonitril
2-Methoxybenzonitril
Formel: C₈H₇NO
Siedepunkt: 73…76 °C (0,1 mmHg)
Melting Pt: 24,5 °C
Dichte: 1,095 g/cm³ (15 °C)
MDL Number: MFCD00001783
CAS-Nummer: 6609-56-9
EINECS: 229-559-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR