2-Nitrobenzonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: o-Nitrobenzonitrile

AS22178G100 AS22178G25
ACTIAS22178G100EA 0 CHF
ACTIAS22178G100 ACTIAS22178G25
2-Nitrobenzonitril
2-Nitrobenzonitril
Formel: C₇H₄N₂O₂
MW: 148,12 g/mol
Siedepunkt: 304 °C (1013 hPa)
Melting Pt: 107…109 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007044
CAS-Nummer: 612-24-8
EINECS: 210-301-0
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR