3,5-Dimethoxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS20128G100 AS20128G25 AS20128G5
ACTIAS20128G100EA 153 CHF
ACTIAS20128G100 ACTIAS20128G25 ACTIAS20128G5
3,5-Dimethoxybenzaldehyd
3,5-Dimethoxybenzaldehyde
Formel: C₉H₁₀O₃
MW: 166.18 g/mol
Siedepunkt: 277
Melting Pt: 48
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003366
CAS-Nummer: 7311-34-4
EINECS: 230-772-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR