3-Hydroxy-4-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS33209G1 AS33209G5
ACTIAS33209G1EA 0 CHF
ACTIAS33209G1 ACTIAS33209G5
3-Hydroxy-4-nitrobenzaldehyd
3-Hydroxy-4-nitrobenzaldehyde
Formel: C₇H₅NO₄
MW: 167,12 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007109
CAS-Nummer: 704-13-2
EINECS: 211-879-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR