3-Methoxy-2-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS32401G1 AS32401G5
ACTIAS32401G1EA 0 CHF
ACTIAS32401G1 ACTIAS32401G5
3-Methoxy-2-nitrobenzaldehyd
3-Methoxy-2-nitrobenzaldehyd
Formel: C₈H₇NO₄
MW: 181.15 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007135
CAS-Nummer: 53055-05-3
EINECS: 258-332-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR