2,6-Dibromanilin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS34560G100
AS34560G5
ACTIAS34560G100EA
0
CHF
ACTIAS34560G100
ACTIAS34560G5
2,6-Dibromanilin
2,6-Dibromanilin
Formel:
C₆H₅Br₂N Siedepunkt: 262 °C (1013 hPa) Melting Pt: 85…86 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00007638 CAS-Nummer: 608-30-0 EINECS: 210-158-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...