3-Brom-5-iodpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS35661G1
AS35661G5
ACTIAS35661G1EA
0
CHF
ACTIAS35661G1
ACTIAS35661G5
3-Brom-5-iodpyridin
3-Bromo-5-iodopyridine
Formel:
C₅H₃BrIN |
MDL Number:
MFCD03086019 CAS-Nummer: 233770-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...