2-Hydroxy-4-(trifluormethyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(Trifluoromethyl)pyridin-2-ol

AS35364G1 AS35364G5
ACTIAS35364G1EA 0 CHF
ACTIAS35364G1 ACTIAS35364G5
2-Hydroxy-4-(trifluormethyl)pyridin
2-Hydroxy-4-(trifluoromethyl)pyridine
Formel: C₆H₄F₃NO
MW: 163.1 g/mol
MDL Number: MFCD00153195
CAS-Nummer: 50650-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR