5-Brom-2-fluor-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-2-fluoro-3-pyridylboronic, 5-Bromo-2-fluoro-3-pyridinylboronic acid, 5-Bromo-2-fluoropyridine-3-boronic acid

AS34962G1 AS34962G10 AS34962G5
ACTIAS34962G1EA 0 CHF
ACTIAS34962G1 ACTIAS34962G10 ACTIAS34962G5
5-Brom-2-fluor-3-pyridinboronsäure
5-Bromo-2-fluoro-3-pyridineboronic acid
Formel: C₅H₄BBrFNO₂
MW: 219.8 g/mol
MDL Number: MFCD04039317
CAS-Nummer: 501435-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR