2-(Trifluormethyl)phenylhydraziniumchlorid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-(Trifluoromethyl)phenylhydrazine hydrochloride, 1-(2-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 2-Hydrazinobenzotrifluoride hydrochloride, (2-(Trifluormethyl)phenyl)hydrazin HCl
AS24147G100
AS24147G25
ACTIAS24147G100EA
316
CHF
ACTIAS24147G100
ACTIAS24147G25
2-(Trifluormethyl)phenylhydraziniumchlorid
2-(Trifluormethyl)phenylhydraziniumchlorid
Formel:
C₇H₈ClF₃N₂ MW: 212,6 g/mol |
MDL Number:
MFCD00102619 CAS-Nummer: 3107-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...