4-Bromphenylhydraziniumchlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride

AS21279G100 AS21279G25
ACTIAS21279G100EA 0 CHF
ACTIAS21279G100 ACTIAS21279G25
4-Bromphenylhydraziniumchlorid
4-Bromphenylhydraziniumchlorid
Formel: C₆H₈BrClN₂
MW: 223,50 g/mol
MDL Number: MFCD00012941
CAS-Nummer: 622-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR