2-Brom-5-chlorbenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS33225G1
AS33225G5
ACTIAS33225G1EA
0
CHF
ACTIAS33225G1
ACTIAS33225G5
2-Brom-5-chlorbenzaldehyd
2-Bromo-5-chlorobenzaldehyde
Formel:
C₇H₄BrClO |
MDL Number:
MFCD00462870 CAS-Nummer: 174265-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...