2-Brom-4-chlorbenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS32486G1
AS32486G5
ACTIAS32486G1EA
0
CHF
ACTIAS32486G1
ACTIAS32486G5
2-Brom-4-chlorbenzaldehyd
2-Bromo-4-chlorobenzaldehyde
Formel:
C₇H₄BrClO MW: 219.46 g/mol |
MDL Number:
MFCD09056792 CAS-Nummer: 84459-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...