Ethylthiooxamat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Thiooxamic acid ethyl ester, Ethyl amino(thioxo)acetate, Ethyl 2-amino-2-thioxoacetate
AS21266G100
AS21266G25
AS21266G5
ACTIAS21266G100EA
0
CHF
ACTIAS21266G100
ACTIAS21266G25
ACTIAS21266G5
Ethylthiooxamat
Ethylthiooxamat
Formel:
C₄H₇NO₂S |
MDL Number:
MFCD00074903 CAS-Nummer: 16982-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...