2-Methyl-3-pyridinol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Hydroxy-2-methylpyridine

AS23402G100 AS23402G25 AS23402G5
ACTIAS23402G100EA 189 CHF
ACTIAS23402G100 ACTIAS23402G25 ACTIAS23402G5
2-Methyl-3-pyridinol
2-Methylpyridin-3-ol
Formel: C₆H₇NO
MW: 109,13 g/mol
MDL Number: MFCD00082538
CAS-Nummer: 1121-25-1
EINECS: 214-327-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR