4-Methyl-2-pyridon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Hydroxy-4-methylpyridine, 4-Methylpyridin-2-ol

AS33697G10 AS33697G100 AS33697G5
ACTIAS33697G10EA 0 CHF
ACTIAS33697G10 ACTIAS33697G100 ACTIAS33697G5
4-Methyl-2-pyridon
4-Methyl-2-pyridon
Formel: C₆H₇NO
MW: 109,13 g/mol
MDL Number: MFCD00134298
CAS-Nummer: 13466-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR