2,6-Difluor-4-methoxyanilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Amino-3,5-difluoroanisole

AS33415G1 AS33415G5
ACTIAS33415G1EA 0 CHF
ACTIAS33415G1 ACTIAS33415G5
2,6-Difluor-4-methoxyanilin
2,6-Difluor-4-methoxyanilin
Formel: C₇H₇F₂NO
MDL Number: MFCD04115922
CAS-Nummer: 151414-47-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR