1-Brom-2,6-difluorbenzol

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

Synonyms: 2-Bromo-1,3-difluorobenzene

AS21623G100 AS21623G25
ACTIAS21623G100EA 246 CHF
ACTIAS21623G100 ACTIAS21623G25
1-Brom-2,6-difluorbenzol
1-Brom-2,6-difluorbenzol
Formel: C₆H₃BrF₂
MW: 192,99 g/mol
Siedepunkt: 61 °C (1013 hPa)
Dichte: 1.71 g/cm³ (20 °C)
MDL Number: MFCD00009894
CAS-Nummer: 64248-56-2
EINECS: 264-750-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR