1-Brom-2,4-dichlorbenzol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 2,4-Dichlorobromobenzene

AS34596G10 AS34596G100 AS34596G5
ACTIAS34596G10EA 0 CHF
ACTIAS34596G10 ACTIAS34596G100 ACTIAS34596G5
1-Brom-2,4-dichlorbenzol
1-Brom-2,4-dichlorbenzol
Formel: C₆H₃BrCl₂
MDL Number: MFCD00018333
CAS-Nummer: 1193-72-2
EINECS: 214-778-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR