2-Brombenzonitril
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
o-Bromobenzonitrile
AS20030G100
AS20030G25
ACTIAS20030G100EA
0
CHF
ACTIAS20030G100
ACTIAS20030G25
2-Brombenzonitril
2-Brombenzonitril
Formel:
C₇H₄BrN MW: 182,02 g/mol Siedepunkt: 252 °C (1013 hPa) Melting Pt: 51…52 °C Dichte: 1.496 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00001772 CAS-Nummer: 2042-37-7 EINECS: 218-045-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...