2-Fluor-3-methoxybenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS30965G1
AS30965G5
ACTIAS30965G1EA
0
CHF
ACTIAS30965G1
ACTIAS30965G5
2-Fluor-3-methoxybenzoesäure
2-Fluoro-3-methoxybenzoic acid
Formel:
C₈H₇FO₃ MW: 170.14 g/mol |
MDL Number:
MFCD02179621 CAS-Nummer: 137654-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...