2-Brom-6-methoxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS32342G1 AS32342G5
ACTIAS32342G1EA 0 CHF
ACTIAS32342G1 ACTIAS32342G5
2-Brom-6-methoxybenzoesäure
2-Bromo-6-methoxybenzoic acid
Formel: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD00800732
CAS-Nummer: 31786-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR