(1S,2S)-(-)-1,2-Diphenylethylendiamin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (1S,2S)-(-)-1,2-Diphenylethylenediamine, (1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, (S,S)-DPEN

AS33768G1 AS33768G10 AS33768G5
ACTIAS33768G1EA 0 CHF
ACTIAS33768G1 ACTIAS33768G10 ACTIAS33768G5
(1S,2S)-(-)-1,2-Diphenylethylendiamin
(1S,2S)-(-)-1,2-Diphenylethylendiamin
Formel: C₁₄H₁₆N₂
MW: 212.29 g/mol
MDL Number: MFCD00082751
CAS-Nummer: 29841-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR