(3-Fluor-4-hydroxyphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

AS35212G1 AS35212G10 AS35212G5
ACTIAS35212G1EA 0 CHF
ACTIAS35212G1 ACTIAS35212G10 ACTIAS35212G5
(3-Fluor-4-hydroxyphenyl)essigsäure
(3-Fluor-4-hydroxyphenyl)essigsäure
Formel: C₈H₇FO₃
MDL Number: MFCD00004348
CAS-Nummer: 458-09-3
EINECS: 207-275-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR