4-Methoxyanthranilsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-4-methoxybenzoic acid

AS31288G1 AS31288G5
ACTIAS31288G1EA 0 CHF
ACTIAS31288G1 ACTIAS31288G5
4-Methoxyanthranilsäure
4-Methoxyanthranilic acid
Formel: C₈H₉NO₃
MW: 167,16 g/mol
MDL Number: MFCD00667729
CAS-Nummer: 4294-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR