2-Chlor-4-methoxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-p-anisic acid

AS32316G1 AS32316G10 AS32316G5
ACTIAS32316G1EA 0 CHF
ACTIAS32316G1 ACTIAS32316G10 ACTIAS32316G5
2-Chlor-4-methoxybenzoesäure
2-Chloro-4-methoxybenzoic acid
Formel: C₈H₇ClO₃
MDL Number: MFCD00085943
CAS-Nummer: 21971-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR