2-Brom-4-fluor-6-methylanilin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Bromo-4-fluoro-o-toluidine
AS25355G100
AS25355G25
AS25355G5
ACTIAS25355G100EA
0
CHF
ACTIAS25355G100
ACTIAS25355G25
ACTIAS25355G5
2-Brom-4-fluor-6-methylanilin
2-Brom-4-fluor-6-methylanilin
Formel:
C₇H₇BrFN MW: 204,04 g/mol |
MDL Number:
MFCD01861196 CAS-Nummer: 202865-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...