Kalium(brommethyl)trifluoroborat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (Bromomethyl)trifluoroboric acid potassium salt, Potassium (bromomethyl)trifluoroborate

AS50355G1 AS50355G5
ACTIAS50355G1EA 0 CHF
ACTIAS50355G1 ACTIAS50355G5
Kalium(brommethyl)trifluoroborat
Kalium(brommethyl)trifluoroborat
Formel: CH₂BBrF₃K
MDL Number: MFCD09265154
CAS-Nummer: 888711-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR