2'-Trifluormethoxyacetophenon
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-[2-(Trifluoromethoxy)phenyl]-1-ethanone, 1-(2-(Trifluoromethoxy)phenyl)ethanone
AS24738G100
AS24738G25
AS24738G5
ACTIAS24738G100EA
414
CHF
ACTIAS24738G100
ACTIAS24738G25
ACTIAS24738G5
2'-Trifluormethoxyacetophenon
2'-Trifluormethoxyacetophenon
Formel:
C₉H₇F₃O₂ MW: 204,15 g/mol |
MDL Number:
MFCD00236320 CAS-Nummer: 220227-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...