2-Mercaptobenzimidazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol

AS34260G10 AS34260G100 AS34260G5
ACTIAS34260G10EA 0 CHF
ACTIAS34260G10 ACTIAS34260G100 ACTIAS34260G5
2-Mercaptobenzimidazol
2-Mercaptobenzimidazole
Formel: C₇H₆N₂S
Siedepunkt: 376 °C (1013 hPa)
Melting Pt: 154 °C
Dichte: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00466107
CAS-Nummer: 583-39-1
EINECS: 209-502-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR