2-Hydroxy-5-nitro-6-picolin
Lieferant: ACTIVATE SCIENTIFIC
AS40246G100
AS40246G5
ACTIAS40246G100EA
0
CHF
ACTIAS40246G100
ACTIAS40246G5
2-Hydroxy-5-nitro-6-picolin
2-Hydroxy-5-nitro-6-picolin
Formel:
C₆H₆N₂O₃ |
MDL Number:
MFCD00092010 CAS-Nummer: 28489-45-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...