2-Hydroxy-3-nitro-6-picolin

Lieferant: ACTIVATE SCIENTIFIC

AS7161G1 AS7161G5
ACTIAS7161G1EA 0 CHF
ACTIAS7161G1 ACTIAS7161G5
2-Hydroxy-3-nitro-6-picolin
2-Hydroxy-3-nitro-6-picolin
Formel: C₆H₆N₂O₃
MDL Number: MFCD03001414
CAS-Nummer: 39745-39-6
EINECS: 244-647-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR