2-Hydroxy-3-nitro-4-picolin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Methyl-3-nitro-2-pyridone, 2-Hydroxy-4-methyl-3-nitropyridine
AS31792G1
AS31792G10
AS31792G5
ACTIAS31792G1EA
0
CHF
ACTIAS31792G1
ACTIAS31792G10
ACTIAS31792G5
2-Hydroxy-3-nitro-4-picolin
2-Hydroxy-3-nitro-4-picolin
Formel:
C₆H₆N₂O₃ MW: 154,13 g/mol Melting Pt: 234…235 °C |
MDL Number:
MFCD00010689 CAS-Nummer: 21901-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...