2-Amino-5-nitro-6-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Amino-2-methyl-3-nitropyridine, 2-Amino-5-nitro-6-methylpyridine, 2-Amino-6-methyl-5-nitropyridine, 6-Amino-3-nitro-2-picoline, 6-Methyl-5-nitro-2-pyridylamine

AS7233G1 AS7233G5
ACTIAS7233G1EA 0 CHF
ACTIAS7233G1 ACTIAS7233G5
2-Amino-5-nitro-6-picolin
2-Amino-5-nitro-6-picoline
Formel: C₆H₇N₃O₂
MW: 153,14 g/mol
MDL Number: MFCD00053582
CAS-Nummer: 22280-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR