6-Nitro-m-toluidin
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
Synonyms:
6-Nitro-3-methylaniline, 3-Amino-4-nitrotoluene, 5-Methyl-2-nitroaniline
AS34959G1
AS34959G10
AS34959G5
ACTIAS34959G1EA
0
CHF
ACTIAS34959G1
ACTIAS34959G10
ACTIAS34959G5
6-Nitro-m-toluidin
6-Nitro-m-toluidin
Formel:
C₇H₈N₂O₂ MW: 152,15 g/mol Melting Pt: 112 °C |
MDL Number:
MFCD00007814 CAS-Nummer: 578-46-1 EINECS: 209-423-7 UN: 2660 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...