4-Iod-3-nitrobenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS34861G1
AS34861G10
AS34861G5
ACTIAS34861G1EA
0
CHF
ACTIAS34861G1
ACTIAS34861G10
ACTIAS34861G5
4-Iod-3-nitrobenzoesäure
4-Iodo-3-nitrobenzoic acid
Formel:
C₇H₄INO₄ MW: 293.02 g/mol |
MDL Number:
MFCD00502399 CAS-Nummer: 35674-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...