2-Iod-1-methylimidazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Iodo-1-methyl-1H-imidazole

AS51310G1 AS51310G5
ACTIAS51310G1EA 0 CHF
ACTIAS51310G1 ACTIAS51310G5
2-Iod-1-methylimidazol
2-Iodo-1-methylimidazole
Formel: C₄H₅IN₂
MW: 208 g/mol
MDL Number: MFCD00661606
CAS-Nummer: 37067-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR