2,5-Diiod-1-methylimidazol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2,5-Diiodo-1-methyl-1H-imidazole
AS51186G1
AS51186G5
ACTIAS51186G1EA
0
CHF
ACTIAS51186G1
ACTIAS51186G5
2,5-Diiod-1-methylimidazol
2,5-Diiodo-1-methylimidazole
Formel:
C₄H₄I₂N₂ MW: 333,9 g/mol |
MDL Number:
MFCD02179540 CAS-Nummer: 86026-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...