2,4-Difluorbenzolsulfonamid
Lieferant: ACTIVATE SCIENTIFIC
AS34950G1
AS34950G10
AS34950G5
ACTIAS34950G1EA
0
CHF
ACTIAS34950G1
ACTIAS34950G10
ACTIAS34950G5
2,4-Difluorbenzolsulfonamid
2,4-Difluorbenzolsulfonamid
Formel:
C₆H₅F₂NO₂S MW: 193,17 g/mol |
MDL Number:
MFCD01320743 CAS-Nummer: 13656-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...