2,3-Difluor-4-nitroanisol
Lieferant: ACTIVATE SCIENTIFIC
AS34992G1
AS34992G5
ACTIAS34992G1EA
0
CHF
ACTIAS34992G1
ACTIAS34992G5
2,3-Difluor-4-nitroanisol
2,3-Difluor-4-nitroanisol
Formel:
C₇H₅F₂NO₃ MW: 189.12 g/mol |
MDL Number:
MFCD07995055 CAS-Nummer: 66684-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...