2-Fluor-3-nitro-6-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-3-nitro-6-methylpyridine, 2-Fluoro-6-methyl-3-nitropyridine

AS33592G1 AS33592G5
ACTIAS33592G1EA 0 CHF
ACTIAS33592G1 ACTIAS33592G5
2-Fluor-3-nitro-6-picolin
2-Fluor-3-nitro-6-picolin
Formel: C₆H₅FN₂O₂
MW: 156,12 g/mol
MDL Number: MFCD03095067
CAS-Nummer: 19346-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR