3,4-Difluor-o-phenylendiamin ≥95%

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,4-Difluoro-1,2-phenylenediamine, 3,4-Difluorobenzene-1,2-diamine, 3,4-Difluoro-o-phenylenediamine

AS21952G1 AS21952G5
ACTIAS21952G1EA 0 CHF
ACTIAS21952G1 ACTIAS21952G5
3,4-Difluor-o-phenylendiamin ≥95%
3,4-Difluor-o-phenylendiamin
Formel: C₆H₆F₂N₂
MW: 144,12 g/mol
MDL Number: MFCD00153128
CAS-Nummer: 153505-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR