2-Fluor-4-iod-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-4-iodo-5-picoline

AS7174G1 AS7174G5
ACTIAS7174G1EA 0 CHF
ACTIAS7174G1 ACTIAS7174G5
2-Fluor-4-iod-5-methylpyridin
2-Fluoro-4-iodo-5-methylpyridine
Formel: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD03095297
CAS-Nummer: 153034-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR