2-Fluor-4-iod-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-4-iodo-3-picoline

AS31775G1 AS31775G5
ACTIAS31775G1EA 0 CHF
ACTIAS31775G1 ACTIAS31775G5
2-Fluor-4-iod-3-methylpyridin
2-Fluoro-4-iodo-3-methylpyridine
Formel: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD03095289
CAS-Nummer: 153034-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR