2-Fluor-3-iod-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-3-iodo-5-picoline

AS33711G1 AS33711G5
ACTIAS33711G1EA 0 CHF
ACTIAS33711G1 ACTIAS33711G5
2-Fluor-3-iod-5-methylpyridin
2-Fluoro-3-iodo-5-methylpyridine
Formel: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD03095303
CAS-Nummer: 153034-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR