Bis(pentafluorphenyl)carbonat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Bis(pentafluorophenyl) Carbonate
AS24725G25
AS24725G5
ACTIAS24725G25EA
120
CHF
ACTIAS24725G25
ACTIAS24725G5
Bis(pentafluorphenyl)carbonat
Bis(pentafluorphenyl)carbonat
Formel:
C₁₃F₁₀O₃ MW: 394,13 g/mol |
MDL Number:
MFCD00368353 CAS-Nummer: 59483-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...